Chitin

Supplier: TCI America

C0072-250G C0072-25G
TCC0072-250GEA 109.51 CAD
TCC0072-250G TCC0072-025G
Chitin
Chitin

CAS Number: 1398-61-4
MDL Number: MFCD00466914
Molecular Formula: C30H50N4O19
Form: Crystal
Color: White

Formula: C₃₀H₅₀N₄O₁₉
MW: 233.26 g/mol
MDL Number: MFCD00466914
CAS Number: 1398-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR