(1R)-Camphor oxime ≥98.0%

Supplier: TCI America

C0013-25G C0013-1G C0013-5G
TCC0013-025GEA 411.59 CAD
TCC0013-025G TCC0013-001G 76219-650
(1R)-Camphor oxime ≥98.0%
(1R)-Camphor oxime

CAS Number: 2792-42-9
MDL Number: MFCD00046298
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119

Formula: C₁₀H₁₇NO
Melting Pt: 119 °C
MDL Number: MFCD00046298
CAS Number: 2792-42-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4d8cf8a81fca57091b0a4636c85e1629f749f80&vid=b4d8cf8a81fca57091b0a4636c85e1629f749f80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR