Calcein (mixture of isomers) for fluorometric determination of Ca
Supplier: TCI America
Synonyms:
Fluorexon (mixture of isomers), Fluorescein Bis(methyliminodiacetic Acid) (mixture of isomers), N,N'-[(3',6'-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthene]-ar',ar'-diyl)bis(methylene)]bis[N-(carboxymethyl)glycine], Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein (mixture of isomers), Calcein
[for Fluorometric Determination of Ca]
CAS Number: 154071-48-4
MDL Number: MFCD00005049
Molecular Formula: C30H26N2O13
Molecular Weight: 622.54
Form: Crystal
Color: Reddish Yellow
Form: Crystal
Color: Reddish Yellow
Formula:
C₃₀H₂₆N₂O₁₃ MW: 622.53 g/mol |
MDL Number:
MFCD00005049 CAS Number: 154071-48-4 EINECS: 215-957-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...