cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
cis-3-(Boc-amino)cyclohexanecarboxylic Acid, cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, cis-3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid
B3518-1G
B3518-5G
TCB3518-1GEA
105.86
CAD
TCB3518-1G
TCB3518-5G
cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97.0% (by titrimetric analysis)
cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
CAS Number: 222530-33-8
MDL Number: MFCD03453252
Molecular Formula: C12H21NO4
Molecular Weight: 243.30
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 145
Other Product Names / Synonyms: cis-3-(Boc-amino)cyclohexanecarboxylic Acid
Formula:
C₁₂H₂₁NO₄ MW: 243.30 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD03453252 CAS Number: 222530-33-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...