2-(3-Bromophenyl)ethanol ≥98.0%
Supplier: TCI America
B3517-25G
B3517-5G
TCB3517-25GEA
529.3
CAD
TCB3517-25G
TCB3517-5G
2-(3-Bromophenyl)ethanol ≥98.0%
2-(3-Bromophenyl)ethanol
CAS Number: 28229-69-8
MDL Number: MFCD00191857
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 113
Specific Gravity (20/20): 1.48
Other Product Names / Synonyms: 2-(3-Bromophenyl)ethanol, 3-Bromophenethyl Alcohol
Formula:
C₈H₉BrO Boiling Pt: 110 °C Density: 1.48 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00191857 CAS Number: 28229-69-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...