2-(2-Bromophenyl)ethanol ≥98.0%
Supplier: TCI America
B3516-5G
B3516-25G
TCB3516-5GEA
118.5
CAD
TCB3516-5G
TCB3516-25G
2-(2-Bromophenyl)ethanol ≥98.0%
2-(2-Bromophenyl)ethanol
CAS Number: 1074-16-4
MDL Number: MFCD00093566
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 113
Specific Gravity (20/20): 1.49
Other Product Names / Synonyms: 2-(2-Bromophenyl)ethanol, 2-Bromophenethyl Alcohol
Formula:
C₈H₉BrO Boiling Pt: 132 °C Density: 1.49 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00093566 CAS Number: 1074-16-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...