2,6-Bis(2-benzimidazolyl)pyridine ≥98.0% (by HPLC)

Supplier: TCI America

B3510-1G B3510-5G
TCB3510-1GEA 178.54 CAD
TCB3510-1G TCB3510-5G
2,6-Bis(2-benzimidazolyl)pyridine ≥98.0% (by HPLC)
2,6-Bis(2-benzimidazolyl)pyridine

CAS Number: 28020-73-7
MDL Number: MFCD00134500
Molecular Formula: C19H13N5
Molecular Weight: 311.35
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 353
Lambda max.: 325 nm (CH3CN)


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 2,2'-(2,6-Pyridinediyl)bis(benzimidazole)

Formula: C₁₉H₁₃N₅
Melting Pt: 353 °C
MDL Number: MFCD00134500
CAS Number: 28020-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5350acb2bfd0e7b3a7dd16f3f9b1dfc4a01897cf&vid=5350acb2bfd0e7b3a7dd16f3f9b1dfc4a01897cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR