tert-Butyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3504-5G B3504-25G
TCB3504-5GEA 127.98 CAD
TCB3504-5G TCB3504-25G
tert-Butyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)
tert-Butyl 4-aminobenzoate

CAS Number: 18144-47-3
MDL Number: MFCD00665790
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: 4-Aminobenzoic Acid tert-Butyl Ester

Formula: C₁₁H₁₅NO₂
Melting Pt: 110 °C
MDL Number: MFCD00665790
CAS Number: 18144-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca0c1e64d03530ec45d13b144a26db011e5002b8&vid=ca0c1e64d03530ec45d13b144a26db011e5002b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR