2-Bromo-4-nitroimidazole ≥98.0%

Supplier: TCI America

B3503-1G B3503-5G
TCB3503-1GEA 91.64 CAD
TCB3503-1G TCB3503-5G
2-Bromo-4-nitroimidazole ≥98.0%
2-Bromo-4-nitroimidazole

CAS Number: 65902-59-2
MDL Number: MFCD09038915
Molecular Formula: C3H2BrN3O2
Molecular Weight: 191.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 235

Formula: C₃H₂BrN₃O₂
Melting Pt: 235 °C
MDL Number: MFCD09025308
CAS Number: 65902-59-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e933f8fdee0503d4a5debb3469bd51b60a1d653&vid=3e933f8fdee0503d4a5debb3469bd51b60a1d653&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR