1,1'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3501-5G
B3501-1G
TCB3501-5GEA
910.08
CAD
TCB3501-5G
TCB3501-1G
1,1'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene ≥98.0% (by GC, titration analysis)
1,1'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene
CAS Number: 737776-93-1
MDL Number: MFCD06411336
Molecular Formula: C22H32B2FeO4
Molecular Weight: 437.96
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 1,1'-Ferrocenediboronic Acid Bis(pinacol) Ester
Formula:
C₂₂H₃₂B₂FeO₄ Melting Pt: 173 °C |
MDL Number:
MFCD06411336 CAS Number: 737776-93-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...