(6-Bromo-3-pyridinyl)methanol ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol

B3494-5G B3494-1G
TCB3494-5GEA 619.36 CAD
TCB3494-5G TCB3494-1G
(6-Bromo-3-pyridinyl)methanol ≥98.0%
(6-Bromo-3-pyridinyl)methanol

CAS Number: 122306-01-8
MDL Number: MFCD04974508
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 50


Other Product Names / Synonyms: 2-Bromo-5-(hydroxymethyl)pyridine

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 50 °C
MDL Number: MFCD04974508
CAS Number: 122306-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ee029f7aec51c5b619a6548406eac7df720881&vid=31ee029f7aec51c5b619a6548406eac7df720881&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR