(5-Bromo-2-pyridinyl)methanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3492-1G B3492-5G
TCB3492-1GEA 74.42 CAD
TCB3492-1G TCB3492-5G
(5-Bromo-2-pyridinyl)methanol ≥98.0% (by GC, titration analysis)
(5-Bromo-2-pyridinyl)methanol

CAS Number: 88139-91-7
MDL Number: MFCD04035597
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 61


Other Product Names / Synonyms: 5-Bromo-2-(hydroxymethyl)pyridine

Formula: C₆H₆BrNO
Boiling Pt: 120 °C
Melting Pt: 61 °C
MDL Number: MFCD04035597
CAS Number: 88139-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea24bb0b276cc502aabc79ef687214690cc9b352&vid=ea24bb0b276cc502aabc79ef687214690cc9b352&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR