2-Bromo-5-formylpyridine ≥98.0%

Supplier: TCI America

B3491-1G B3491-5G
TCB3491-1GEA 34.76 CAD
TCB3491-1G TCB3491-5G
2-Bromo-5-formylpyridine ≥98.0%
2-Bromo-5-formylpyridine

CAS Number: 149806-06-4
MDL Number: MFCD04115419
Molecular Formula: C6H4BrNO
Molecular Weight: 186.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: 2-Bromo-5-formylpyridine, 6-Bromonicotinaldehyde

Formula: C₆H₄BrNO
Melting Pt: 102 °C
MDL Number: MFCD04115419
CAS Number: 149806-06-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR