3-Bromo-2-nitroanisole ≥98.0%
Supplier: TCI America
B3486-5G
B3486-1G
TCB3486-5GEA
134.3
CAD
TCB3486-5G
TCB3486-1G
3-Bromo-2-nitroanisole ≥98.0%
3-Bromo-2-nitroanisole
CAS Number: 500298-30-6
MDL Number: MFCD11101044
Molecular Formula: C7H6BrNO3
Molecular Weight: 232.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 131
Melting point (°C): 74
Other Product Names / Synonyms: 1-Bromo-3-methoxy-2-nitrobenzene
Formula:
C₇H₆BrNO₃ Boiling Pt: 131 °C Melting Pt: 74 °C |
MDL Number:
MFCD11101044 CAS Number: 500298-30-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...