(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-Tyr(tBu)-OH, N-Boc-O-tert-butyl-L-tyrosine

B3485-5G B3485-1G
TCB3485-5GEA 137.46 CAD
TCB3485-5G TCB3485-1G
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0%
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

CAS Number: 47375-34-8
MDL Number: MFCD00065598
Molecular Formula: C18H27NO5
Molecular Weight: 337.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 117
Specific rotation [a]20/D: -16 deg (C=1, DMF)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-O-tert-butyl-L-tyrosine, Boc-Tyr(tBu)-OH

Formula: C₁₈H₂₇NO₅
MW: 337.4 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00065598
CAS Number: 47375-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0fd1d9e95f04f0a9dddcdc531cc9413f57348b6&vid=b0fd1d9e95f04f0a9dddcdc531cc9413f57348b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR