4-tert-Butyldiphenyl sulfide ≥98.0%
Supplier: TCI America
B3484-100G
B3484-25G
TCB3484-100GEA
575.12
CAD
TCB3484-100G
TCB3484-25G
4-tert-Butyldiphenyl sulfide ≥98.0%
4-tert-Butyldiphenyl sulfide
CAS Number: 85609-03-6
MDL Number: MFCD06200845
Molecular Formula: C16H18S
Molecular Weight: 242.38
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 1.04
Formula:
C₁₆H₁₈S Boiling Pt: 140 °C Density: 1.04 (20 °C) |
MDL Number:
MFCD06200845 CAS Number: 85609-03-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...