Sodium 4-acetamidobenzenesulfinate dihydrate ≥98.0%

Supplier: TCI America

Synonyms: 4-Acetamidobenzenesulfinic Acid Sodium Salt, 4-Acetamidobenzenesulfinic acid Sodium Salt Dihydrate, 4-Acetamidobenzenesulphinic Acid Sodium Salt, Sodium 4-Acetamidobenzenesulphinate Dihydrate, Sodium 4-Acetamidobenzenesulfinate Dihydrate

A0011-5G A0011-25G
TCA0011-005GEA 485.06 CAD
TCA0011-005G TCA0011-025G
Sodium 4-acetamidobenzenesulfinate dihydrate ≥98.0%
Sodium 4-acetamidobenzenesulfinate dihydrate

CAS Number: 15898-43-8
MDL Number: MFCD04113633
Molecular Formula: C8H9NO3S
Molecular Weight: 221.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-Acetamidobenzenesulfinic Acid Sodium Salt

Formula: C₈H₉NO₃S
MW: 221.21 g/mol
MDL Number: MFCD04113633
CAS Number: 15898-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a7b0520cedb850009da84cb0843cd50524a86b9&vid=3a7b0520cedb850009da84cb0843cd50524a86b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR