1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
IPr, 1,3-Bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene
CAS Number: 244187-81-3
MDL Number: MFCD07781991
Molecular Formula: C27H36N2
Molecular Weight: 388.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 217
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, N-Heterocyclic Carbene Ligands ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: IPr
Formula:
C₂₇H₃₆N₂ MW: 388.60 g/mol Melting Pt: 217 °C |
MDL Number:
MFCD07781991 CAS Number: 244187-81-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...