Benzo[1,2-b:4,5-b']dithiophene ≥98.0%

Supplier: TCI America

B3464-5G B3464-1G
TCB3464-5GEA 955.9 CAD
TCB3464-5G TCB3464-1G
Benzo[1,2-b:4,5-b']dithiophene ≥98.0%
Benzo[1,2-b:4,5-b']dithiophene

CAS Number: 267-65-2
Molecular Formula: C10H6S2
Molecular Weight: 190.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 198
Lambda max.: 334 nm (EtOH)

Formula: C₁₀H₆S₂
Melting Pt: 198 °C
CAS Number: 267-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26b98b20f7494e058bd01685f6611739e4fe1cb9&vid=26b98b20f7494e058bd01685f6611739e4fe1cb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR