3-Bromo-9-ethylcarbazole ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-9-ethyl-9H-carbazole

B3459-1G B3459-5G
TCB3459-1GEA 54.51 CAD
TCB3459-1G TCB3459-5G
3-Bromo-9-ethylcarbazole ≥98.0%
3-Bromo-9-ethylcarbazole

CAS Number: 57102-97-3
MDL Number: MFCD00092392
Molecular Formula: C14H12BrN
Molecular Weight: 274.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84

Formula: C₁₄H₁₂BrN
MW: 274.1558 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00092392
CAS Number: 57102-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8970c121a43e5b51a4f78bdac7e2a0f44648b7bd&vid=8970c121a43e5b51a4f78bdac7e2a0f44648b7bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR