1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea ≥98.0%

Supplier: TCI America

B3452-200MG B3452-1G
TCB3452-200MGEA 107.44 CAD
TCB3452-200MG TCB3452-1G
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea ≥98.0%
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea

CAS Number: 1060-92-0
Molecular Formula: C17H8F12N2S
Molecular Weight: 500.31
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 152

Formula: C₁₇H₈F₁₂N₂S
MW: 500.31 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00829878
CAS Number: 1060-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0caa59bec00356e9fd94c380fab7bc6d872427e9&vid=0caa59bec00356e9fd94c380fab7bc6d872427e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR