8-Bromo-2-methylquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3444-1G B3444-5G
TCB3444-1GEA 120.08 CAD
TCB3444-1G TCB3444-5G
8-Bromo-2-methylquinoline ≥98.0% (by GC, titration analysis)
8-Bromo-2-methylquinoline

CAS Number: 61047-43-6
MDL Number: MFCD04966997
Molecular Formula: C10H8BrN
Molecular Weight: 222.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 68


Other Product Names / Synonyms: 8-Bromoquinaldine

Formula: C₁₀H₈BrN
MW: 222.0812 g/mol
Boiling Pt: 132 °C
Melting Pt: 68 °C
MDL Number: MFCD04966997
CAS Number: 61047-43-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf6f9fec03db4494bc41656b19e3a9753fd4d226&vid=bf6f9fec03db4494bc41656b19e3a9753fd4d226&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR