p-Tolyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic acid p-Cresyl ester, Acetic acid p-Tolyl ester, p-Tolyl Acetate, p-Cresyl acetate

A0670-25G A0670-500G
TCA0670-025GEA 43.29 CAD
TCA0670-025G TCA0670-500G
p-Tolyl acetate ≥98.0%
p-Tolyl acetate

CAS Number: 140-39-6
MDL Number: MFCD00008703
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 95
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Acetic Acid p-Cresyl Ester, Acetic Acid p-Tolyl Ester, p-Cresyl Acetate

Formula: C₉H₁₀O₂
MW: 150.1745 g/mol
Boiling Pt: 214 °C
Density: 1.05 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00008703
CAS Number: 140-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=522d44206c14af254f1765123f0625894fcc12ab&vid=522d44206c14af254f1765123f0625894fcc12ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR