4-Bromo-N-methylphthalimide ≥98.0%

Supplier: TCI America

B3443-5G B3443-25G
TCB3443-5GEA 219.62 CAD
TCB3443-5G TCB3443-25G
4-Bromo-N-methylphthalimide ≥98.0%
4-Bromo-N-methylphthalimide

CAS Number: 90224-73-0
Molecular Formula: C9H6BrNO2
Molecular Weight: 240.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150

Formula: C₉H₆BrNO₂
Melting Pt: 150 °C
CAS Number: 90224-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ac4bb12468e0ca3b857cd06f0ac32303536faf6&vid=9ac4bb12468e0ca3b857cd06f0ac32303536faf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR