2,5-Bis(4-biphenylyl)thiophene

Supplier: TCI America

B3441-1G B3441-5G
TCB3441-1GEA 233.05 CAD
TCB3441-1G TCB3441-5G
2,5-Bis(4-biphenylyl)thiophene
2,5-Bis(4-biphenylyl)thiophene

CAS Number: 56316-86-0
Molecular Formula: C28H20S
Molecular Weight: 388.53
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 337
Lambda max.: 356 nm (CH2Cl2)


Other Product Names / Synonyms: BP1T

Formula: C₂₈H₂₀S
Melting Pt: 337 °C
CAS Number: 56316-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4cb2791b0384e99ae04d1b028439b12758aca81&vid=b4cb2791b0384e99ae04d1b028439b12758aca81&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR