trans-4-(tert-Butyldiphenylsilyloxy)-L-proline ≥98.0%

Supplier: TCI America

Synonyms: (2S,4R)-4-(tert-Butyldiphenylsilyloxy)pyrrolidine-2-carboxylic Acid

B3440-1G B3440-5G
TCB3440-1GEA 285.98 CAD
TCB3440-1G TCB3440-5G
trans-4-(tert-Butyldiphenylsilyloxy)-L-proline ≥98.0%
trans-4-(tert-Butyldiphenylsilyloxy)-L-proline

CAS Number: 259212-61-8
Molecular Formula: C21H27NO3Si
Molecular Weight: 369.54
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198
Specific rotation [a]20/D: -36 deg (C=0.8, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (2S,4R)-4-(tert-Butyldiphenylsilyloxy)pyrrolidine-2-carboxylic Acid

Formula: C₂₁H₂₇NO₃Si
MW: 369.54 g/mol
Melting Pt: 198 °C
CAS Number: 259212-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3cab087845505003e15c007dd5a0dec04f1039c&vid=c3cab087845505003e15c007dd5a0dec04f1039c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR