1,3-Bis(3-hydroxy-3-pentyl)benzene ≥90.0%

Supplier: TCI America

Synonyms: 1,3-Bis(3-hydroxy-3-amyl)benzene, 1,3-Bis(1-ethyl-1-hydroxypropyl)benzene, 3,3'-(m-Phenylene)di-3-pentanol, alpha,alpha,alpha',alpha'-Tetraethyl-1,3-benzenedimethanol

B3436-5G B3436-25G
TCB3436-5GEA 271.13 CAD
TCB3436-5G TCB3436-25G
1,3-Bis(3-hydroxy-3-pentyl)benzene ≥90.0%
1,3-Bis(3-hydroxy-3-pentyl)benzene

CAS Number: 676465-94-4
Molecular Formula: C16H26O2
Molecular Weight: 250.38
Purity/Analysis Method: >90.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 1,3-Bis(3-hydroxy-3-amyl)benzene, 1,3-Bis(1-ethyl-1-hydroxypropyl)benzene, 3,3'-(m-Phenylene)di-3-pentanol, alpha,alpha,alpha',alpha'-Tetraethyl-1,3-benzenedimethanol

Formula: C₁₆H₂₆O₂
MW: 250.38 g/mol
MDL Number: MFCD14708166
CAS Number: 676465-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18555b803453dad24dabdfef4ec4dd392ca91602&vid=18555b803453dad24dabdfef4ec4dd392ca91602&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR