Bis(2-hydroxyethyl)terephthalate ≥85.0% (by HPLC)

Supplier: TCI America

Synonyms: BHET, Terephthalic Acid Bis(2-hydroxyethyl) Ester, Bis(2-hydroxyethyl) terephthalate

B3429-500G B3429-25G
TCB3429-500GEA 232.89 CAD
TCB3429-500G TCB3429-25G
Bis(2-hydroxyethyl)terephthalate ≥85.0% (by HPLC)
Bis(2-hydroxyethyl)terephthalate

CAS Number: 959-26-2
MDL Number: MFCD00081066
Molecular Formula: C12H14O6
Molecular Weight: 254.24
Purity/Analysis Method: >85.0% (HPLC)
Form: Crystal
Melting point (°C): 106
Flash Point (°C): 200


Other Product Names / Synonyms: Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET

Formula: C₁₂H₁₄O₆
MW: 254.24 g/mol
Melting Pt: 106 °C
Flash Pt: 200 °C
MDL Number: MFCD00081066
CAS Number: 959-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afc162325f847136a83ba13c92c8e62633aa719e&vid=afc162325f847136a83ba13c92c8e62633aa719e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR