1-Butyl-1-methylpiperidinium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B3424-5G B3424-25G
TCB3424-5GEA 143.78 CAD
TCB3424-5G TCB3424-25G
1-Butyl-1-methylpiperidinium bromide ≥97.0% (by titrimetric analysis)
1-Butyl-1-methylpiperidinium bromide

CAS Number: 94280-72-5
Molecular Formula: C10H22BrN
Molecular Weight: 236.20
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 230

Formula: C₁₀H₂₂BrN
Melting Pt: 230 °C
MDL Number: MFCD11870112
CAS Number: 94280-72-5

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR