o-Tolyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic acid o-Tolyl ester, Acetic acid o-Cresyl ester, o-Cresyl acetate

A0669-25G A0669-500G
TCA0669-025GEA 91.64 CAD
TCA0669-025G TCA0669-500G
o-Tolyl acetate ≥98.0%
o-Tolyl acetate

CAS Number: 533-18-6
MDL Number: MFCD00053704
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Acetic Acid o-Cresyl Ester, Acetic Acid o-Tolyl Ester, o-Cresyl Acetate

Formula: C₉H₁₀O₂
MW: 150.1700 g/mol
Boiling Pt: 208 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00053704
CAS Number: 533-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeb4848eca0332a4ceb9d01e39f5c64c71276b48&vid=aeb4848eca0332a4ceb9d01e39f5c64c71276b48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR