1,4-Butanediolbis(thioglycolate) ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,4-Bis(mercaptoacetoxy)butane

B3423-25G B3423-500G
TCB3423-25GEA 72.68 CAD
TCB3423-25G TCB3423-500G
1,4-Butanediolbis(thioglycolate) ≥95.0% (by GC, titration analysis)
1,4-Butanediolbis(thioglycolate)

CAS Number: 10193-95-0
MDL Number: MFCD00266655
Molecular Formula: C8H14O4S2
Molecular Weight: 238.32
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.24


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Bis(mercaptoacetoxy)butane

Formula: C₈H₁₄O₄S₂
MW: 238.32 g/mol
Density: 1.24 (20 °C)
MDL Number: MFCD00266655
CAS Number: 10193-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a286c646c8abbf1dc7ae9591dcd394a813263de&vid=4a286c646c8abbf1dc7ae9591dcd394a813263de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR