N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester ≥98.0%

Supplier: TCI America

Synonyms: Boc-D-Pro-OSu, N-Boc-D-proline Succinimidyl Ester

B3414-1G B3414-5G
TCB3414-1GEA 45.82 CAD
TCB3414-1G TCB3414-5G
N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester ≥98.0%
N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester

CAS Number: 102185-34-2
MDL Number: MFCD00069687
Molecular Formula: C14H20N2O6
Molecular Weight: 312.32
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Specific rotation [a]20/D: 56 deg (C=1, Dioxane)
Storage Temperature: <0°C


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Boc-D-proline Succinimidyl Ester, Boc-D-Pro-OSu

Formula: C₁₄H₂₀N₂O₆
MW: 312.32 g/mol
MDL Number: MFCD00069687
CAS Number: 102185-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=474393651c181ac5348d94426fc4d10a78a5fd66&vid=474393651c181ac5348d94426fc4d10a78a5fd66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR