(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: [3S-(3α,4αβ,8αβ)]-N-(tert-Butyl)decahydroisoquinoline-3-carboxamide, (3S,4aS,8aS)-N-(tert-Butyl)decahydroisoquinoline-3-carboxamide

B3410-25G B3410-5G
TCB3410-25GEA 285.98 CAD
TCB3410-25G TCB3410-5G
(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide ≥97.0% (by GC, titration analysis)
(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide

CAS Number: 136465-81-1
MDL Number: MFCD01313226
Molecular Formula: C14H26N2O
Molecular Weight: 238.38
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 115
Specific rotation [a]20/D: -73 deg (C=1, MeOH)

Formula: C₁₄H₂₆N₂O
MW: 238.38 g/mol
Melting Pt: 115 °C
MDL Number: MFCD01313226
CAS Number: 136465-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b4bcccbe56e292c0cc3b275a37cbbdbdfbef327&vid=5b4bcccbe56e292c0cc3b275a37cbbdbdfbef327&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR