2-Benzylacrylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3409-25G
B3409-5G
TCB3409-25GEA
52.14
CAD
TCB3409-25G
TCB3409-5G
2-Benzylacrylic acid ≥98.0% (by GC, titration analysis)
2-Benzylacrylic acid
CAS Number: 5669-19-2
MDL Number: MFCD00854174
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 70
Other Product Names / Synonyms: 2-Methylene-3-phenylpropionic Acid
Formula:
C₁₀H₁₀O₂ Melting Pt: 70 °C |
MDL Number:
MFCD00854174 CAS Number: 5669-19-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...