Bicyclohexane-4,4'-dione monoethylene ketal ≥98.0%

Supplier: TCI America

Synonyms: 8-(4-Oxocyclohexyl)-1,4-dioxaspiro[4.5]decane, Bicyclohexane-4,4-dione Monoethylene Ketal

B3405-25G B3405-5G
TCB3405-25GEA 155.31 CAD
TCB3405-25G TCB3405-5G
Bicyclohexane-4,4'-dione monoethylene ketal ≥98.0%
Bicyclohexane-4,4'-dione monoethylene ketal

CAS Number: 56309-94-5
MDL Number: MFCD00809704
Molecular Formula: C14H22O3
Molecular Weight: 238.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: 8-(4-Oxocyclohexyl)-1,4-dioxaspiro[4.5]decane

Formula: C₁₄H₂₂O₃
MW: 238.33 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00809704
CAS Number: 56309-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abd7658bcaf195d89774f363355c55b4a6fef672&vid=abd7658bcaf195d89774f363355c55b4a6fef672&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR