2-Bromo-3-methoxynaphthalene ≥97.0%

Supplier: TCI America

B3403-1G B3403-5G
TCB3403-1GEA 134.3 CAD
TCB3403-1G TCB3403-5G
2-Bromo-3-methoxynaphthalene ≥97.0%
2-Bromo-3-methoxynaphthalene

CAS Number: 68251-77-4
Molecular Formula: C11H9BrO
Molecular Weight: 237.10
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 75

Formula: C₁₁H₉BrO
MW: 237.0926 g/mol
Melting Pt: 75 °C
MDL Number: MFCD11870110
CAS Number: 68251-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e79f0ea13704b67632a25d044cb1dc9bf85c8fba&vid=e79f0ea13704b67632a25d044cb1dc9bf85c8fba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR