2,6-Bis(hydroxymethyl)naphthalene ≥98.0%

Supplier: TCI America

B3399-1G B3399-5G
TCB3399-1GEA 152 CAD
TCB3399-1G TCB3399-5G
2,6-Bis(hydroxymethyl)naphthalene ≥98.0%
2,6-Bis(hydroxymethyl)naphthalene

CAS Number: 5859-93-8
Molecular Formula: C12H12O2
Molecular Weight: 188.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 173

Formula: C₁₂H₁₂O₂
Melting Pt: 173 °C
MDL Number: MFCD11870109
CAS Number: 5859-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9281a4f20e19caa222105168b64a6b046553b317&vid=9281a4f20e19caa222105168b64a6b046553b317&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR