O-tert-Butyl-L-threonine ≥98.0%

Supplier: TCI America

Synonyms: (2S,3R)-2-Amino-3-(tert-butoxy)butanoic acid, H-Thr(tBu)-OH

B3398-1G B3398-5G
TCB3398-1GEA 39.5 CAD
TCB3398-1G TCB3398-5G
O-tert-Butyl-L-threonine ≥98.0%
O-tert-Butyl-L-threonine

CAS Number: 4378-13-6
MDL Number: MFCD00037766
Molecular Formula: C8H17NO3
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 260
Specific rotation [a]20/D: -43 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-Thr(tBu)-OH

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Melting Pt: 260 °C
MDL Number: MFCD00037766
CAS Number: 4378-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=573ede6bc74cb4dd17e1ef8e0bb1feec09d1ffe7&vid=573ede6bc74cb4dd17e1ef8e0bb1feec09d1ffe7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR