4-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-2-thiophenecarboxylic acid

B3386-25G B3386-5G
TCB3386-25GEA 448.72 CAD
TCB3386-25G TCB3386-5G
4-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-2-thenoic acid

CAS Number: 16694-18-1
MDL Number: MFCD03422294
Molecular Formula: C5H3BrO2S
Molecular Weight: 207.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 122
Lambda max.: 245 nm (EtOH)

Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
Melting Pt: 122 °C
MDL Number: MFCD03422294
CAS Number: 16694-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c05f44564d653c09786d2242041ca1a26975c026&vid=c05f44564d653c09786d2242041ca1a26975c026&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR