trans-2-Butene-1,4-diol ≥95.0%

Supplier: TCI America

Synonyms: (E)-But-2-ene-1,4-diol, trans-1,4-Dihydroxy-2-butene

B3384-1G B3384-5G
TCB3384-1GEA 226.41 CAD
TCB3384-1G TCB3384-5G
trans-2-Butene-1,4-diol ≥95.0%
trans-2-Butene-1,4-diol

CAS Number: 821-11-4
MDL Number: MFCD00063207
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 25


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: trans-1,4-Dihydroxy-2-butene

Formula: C₄H₈O₂
Boiling Pt: 127 °C
Melting Pt: 25 °C
MDL Number: MFCD00063207
CAS Number: 821-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=032571505cc1eccacb31f0cc94632a8ccf065e76&vid=032571505cc1eccacb31f0cc94632a8ccf065e76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR