Butyl-2-methylvalerate ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylpentanoic Acid Butyl Ester, 2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate

B3359-25ML B3359-500ML
TCB3359-25MLEA 50.56 CAD
TCB3359-25ML TCB3359-500ML
Butyl-2-methylvalerate ≥98.0%
Butyl-2-methylvalerate

CAS Number: 6297-41-2
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate, 2-Methylpentanoic Acid Butyl Ester

Formula: C₁₀H₂₀O₂
Density: 0.86 (20 °C)
CAS Number: 6297-41-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4c2a45d705d3b8e88e849bae5d9355ccfdfed7f&vid=d4c2a45d705d3b8e88e849bae5d9355ccfdfed7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR