1-Bromo-4-(trimethylsilyl)benzene ≥97.0%
Supplier: TCI America
B3357-25G
B3357-5G
TCB3357-25GEA
616.2
CAD
TCB3357-25G
TCB3357-5G
1-Bromo-4-(trimethylsilyl)benzene ≥97.0%
1-Bromo-4-(trimethylsilyl)benzene
CAS Number: 6999-03-7
MDL Number: MFCD00094165
Molecular Formula: C9H13BrSi
Molecular Weight: 229.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 96
Specific Gravity (20/20): 1.22
Other Product Names / Synonyms: (4-Bromophenyl)trimethylsilane
Formula:
C₉H₁₃BrSi Boiling Pt: 230 °C Density: 1.22 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00094165 CAS Number: 6999-03-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...