6-Benzyloxypurine 98,0

Supplier: TCI America

B3355-5G B3355-25G
TCB3355-5GEA 34.76 CAD
TCB3355-5G TCB3355-25G
6-Benzyloxypurine 98,0
6-Benzyloxypurine

CAS Number: 57500-07-9
MDL Number: MFCD00213666
Molecular Formula: C12H10N4O
Molecular Weight: 226.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 171

Formula: C₁₂H₁₀N₄O
MW: 226.24 g/mol
Melting Pt: 171 °C
MDL Number: MFCD00213666
CAS Number: 57500-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f75d20989008787bc16f6b13f9b0efd411af58b&vid=8f75d20989008787bc16f6b13f9b0efd411af58b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR