[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B3351-200MG
B3351-1G
TCB3351-200MGEA
94.8
CAD
TCB3351-200MG
TCB3351-1G
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) ≥98.0% (by titrimetric analysis)
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I)
CAS Number: 920739-11-3
Molecular Formula: C42H47CuN2O2
Molecular Weight: 675.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₄₂H₄₇CuN₂O₂ MW: 675.40 g/mol |
CAS Number:
920739-11-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...