Bisdemethoxycurcumin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3347-25G B3347-5G
TCB3347-25GEA 480.32 CAD
TCB3347-25G TCB3347-5G
Bisdemethoxycurcumin ≥98.0% (by HPLC, titration analysis)
Bisdemethoxycurcumin

CAS Number: 24939-16-0
MDL Number: MFCD03419284
Molecular Formula: C19H16O4
Molecular Weight: 308.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 224


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Didemethoxycurcumin

Formula: C₁₉H₁₆O₄
Melting Pt: 224 °C
MDL Number: MFCD03419284
CAS Number: 24939-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=512f0bb01b8f047875e74fc017aab0415f1c8819&vid=512f0bb01b8f047875e74fc017aab0415f1c8819&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR