O-tert-Butyl-L-threonine-tert-butyl ester ≥96.0% (by titrimetric analysis)

Supplier: TCI America

B3340-25G B3340-5G
TCB3340-25GEA 355.5 CAD
TCB3340-25G TCB3340-5G
O-tert-Butyl-L-threonine-tert-butyl ester ≥96.0% (by titrimetric analysis)
O-tert-Butyl-L-threonine-tert-butyl ester

CAS Number: 5854-78-4
MDL Number: MFCD00077109
Molecular Formula: C12H25NO3
Molecular Weight: 231.34
Purity/Analysis Method: >96.0% (T)
Form: Clear Liquid
Boiling point (°C): 70
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: 2.2 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Thr(tBu)-OtBu

Formula: C₁₂H₂₅NO₃
Boiling Pt: 70 °C
Density: 0.94 (20 °C)
MDL Number: MFCD00077109
CAS Number: 5854-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=183e4757f01c521b2e557bdec13dd1b950582aed&vid=da108bdb099f13976afd4d576fb722e60749233e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR