[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid

B3338-1G B3338-5G
TCB3338-1GEA 33.18 CAD
TCB3338-1G TCB3338-5G
[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98.0%
[(tert-Butoxycarbonyl)aminooxy]acetic acid

CAS Number: 42989-85-5
MDL Number: MFCD01632027
Molecular Formula: C7H13NO5
Molecular Weight: 191.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 115


Other Product Names / Synonyms: (Boc-aminooxy)acetic Acid, N-Boc-(carboxymethoxy)amine

Formula: C₇H₁₃NO₅
MW: 191.18 g/mol
Melting Pt: 115 °C
MDL Number: MFCD01632027
CAS Number: 42989-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37e86091fdd73e1d8ef5bdfa8be6b3cbce87db35&vid=37e86091fdd73e1d8ef5bdfa8be6b3cbce87db35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR