7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B3333-5G B3333-1G
TCB3333-5GEA 186.76 CAD
TCB3333-5G TCB3333-1G
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone ≥98.0% (by HPLC, total nitrogen)
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone

CAS Number: 129722-34-5
MDL Number: MFCD06658540
Molecular Formula: C13H16BrNO2
Molecular Weight: 298.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 112


Other Product Names / Synonyms: 7-(4-Bromobutoxy)-1,2,3,4-tetrahydro-2-oxoquinoline

Formula: C₁₃H₁₆BrNO₂
Melting Pt: 112 °C
MDL Number: MFCD06658540
CAS Number: 129722-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe53784801e495c2dd8f923d4d2fea68d033806c&vid=fe53784801e495c2dd8f923d4d2fea68d033806c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR