5-Benzyloxy-2-bromo-4-methoxybenzoic acid ≥97.0%

Supplier: TCI America

Synonyms: 5-Benzyloxy-2-bromo-p-anisic Acid

B3332-5G B3332-1G
TCB3332-5GEA 537.2 CAD
TCB3332-5G TCB3332-1G
5-Benzyloxy-2-bromo-4-methoxybenzoic acid ≥97.0%
5-Benzyloxy-2-bromo-4-methoxybenzoic acid

CAS Number: 24958-42-7
MDL Number: MFCD03695499
Molecular Formula: C15H13BrO4
Molecular Weight: 337.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 195


Other Product Names / Synonyms: 5-Benzyloxy-2-bromo-p-anisic Acid

Formula: C₁₅H₁₃BrO₄
Melting Pt: 195 °C
MDL Number: MFCD03695499
CAS Number: 24958-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=296692585c57c2130abda5a7b3b53e6c87bfe61b&vid=296692585c57c2130abda5a7b3b53e6c87bfe61b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR