5-Bromo-8-quinolinol ≥96.0%

Supplier: TCI America

Synonyms: 5-Bromoquinolin-8-ol

B3324-5G B3324-25G
TCB3324-5GEA 401.32 CAD
TCB3324-5G TCB3324-25G
5-Bromo-8-quinolinol ≥96.0%
5-Bromo-8-quinolinol

CAS Number: 1198-14-7
MDL Number: MFCD00457011
Molecular Formula: C9H6BrNO
Molecular Weight: 224.06
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 127


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 5-Bromo-8-quinolinol

Formula: C₉H₆BrNO
MW: 224.06
Melting Pt: 127 °C
MDL Number: MFCD00457011
CAS Number: 1198-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f783724f9e0cec859bda1f8f02819997fbced506&vid=f783724f9e0cec859bda1f8f02819997fbced506&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR